3-(4-Aminophenyl)propionic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Aminohydrocinnamic acid, 3-(4-Aminophenyl)propanoic acid

A151460-1G A151460-5G
ALA151460-1GEA 17.01 USD
ALA151460-1G ALA151460-5G
3-(4-Aminophenyl)propionic acid ≥98%
3-(4-Aminophenyl)propionic acid
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 133-137°C
MDL Number: MFCD00017118
CAS Number: 2393-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a437042af10fb1cd307a580d32fc279368faf739&vid=a437042af10fb1cd307a580d32fc279368faf739&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR