4-Amino-4'-dimethylaminoazobenzene ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: N,N-Dimethyl-4,4'-azodianiline

A151441-1G A151441-250MG A151441-5G
ALA151441-1GEA 159.51 USD
ALA151441-1G ALA151441250MG ALA151441-5G
4-Amino-4'-dimethylaminoazobenzene ≥97%
4-Amino-4'-dimethylaminoazobenzene
Formula: C₁₄H₁₆N₄
MW: 240.31 g/mol
Melting Pt: 189 °C
Storage Temperature: Ambient
MDL Number: MFCD00010204
CAS Number: 539-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c166c674dce046d42110a458ab5970e76dc5dc4b&vid=c166c674dce046d42110a458ab5970e76dc5dc4b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR