Pentadecylamine ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Pentadecylamine, 1-Aminopentadecane, Pentadecan-1-amine

A151435-1G A151435-5G
ALA151435-1GEA 30.31 USD
ALA151435-1G ALA151435-5G
Pentadecylamine ≥95% (by GC)
Pentadecylamine
Formula: H₃C(CH₂)₁₃CH₂NH₂
Boiling Pt: 299-301°C
Melting Pt: 35-39°C
Flash Pt: 113 °C
MDL Number: MFCD00038195
CAS Number: 2570-26-5
UN: 2923
ADR: 8, 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d79e4cbaea79b164b09b16b7b53d9f427131251&vid=3d79e4cbaea79b164b09b16b7b53d9f427131251&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR