Tridecylamine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: n-Tridecylamine, Tridecan-1-amine, 1-Aminotridecane

A151434-1G A151434-5G
ALA151434-1GEA 56.91 USD
ALA151434-1G ALA151434-5G
Tridecylamine ≥98%
Tridecylamine

Tridecylamine is generally used in introducing C13 chain to the substrate. Some of the application are: · Synthesis of alkylated 1,2,4-triazoles as bridging ligand in the preparation of polymeric 1-dimensional chains of iron(II) species. · Synthesis of amphiphiles such as N-tridecyl-β-hydroxypropionic acid amide (THPA) and N-(β-hydroxyethyl)tridecanoic acid amide (HETA). · As a ligand in the preparation of palladium-based catalyst, [PdCl2(TDA)2].

Formula: H₃C(CH₂)₁₁CH₂NH₂
MW: 199.38 g/mol
Boiling Pt: 265
Melting Pt: 28.1
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008156
CAS Number: 2869-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8da9880627703109c8140853f490511351dafe8c&vid=8da9880627703109c8140853f490511351dafe8c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR