5'-Amino-2'-methylacetanilide ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-Acetamido-4-methylaniline, N-(5-Amino-2-methylphenyl)acetamide, N3-Acetyl-4-methyl-1,3-phenylenediamine
A151426-1G
A151426-250MG
A151426-5G
ALA151426-1GEA
95.86
USD
ALA151426-1G
ALA151426250MG
ALA151426-5G
5'-Amino-2'-methylacetanilide ≥98%
5'-Amino-2'-methylacetanilide
Formula:
C₉H₁₂N₂O Melting Pt: 144 °C |
MDL Number:
MFCD00047859 CAS Number: 5434-30-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...