3-Amino-4-methylbenzamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151422-5G A151422-25G
ALA151422-5GEA 41.71 USD
ALA151422-5G ALA151422-25G
3-Amino-4-methylbenzamide ≥98%
3-Amino-4-methylbenzamide
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Melting Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00035936
CAS Number: 19406-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00f0bc5a913525d2247440880da3f6ab274997fe&vid=00f0bc5a913525d2247440880da3f6ab274997fe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR