6-Amino-1-methyl-5-(propylamino)uracil ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151405-1G A151405-250MG
ALA151405-1GEA 157.61 USD
ALA151405-1G ALA151405250MG
6-Amino-1-methyl-5-(propylamino)uracil ≥98%
6-Amino-1-methyl-5-(propylamino)uracil
Formula: C₈H₁₄N₄O₂
MW: 198.23 g/mol
Melting Pt: 220 °C
MDL Number: MFCD01074825
CAS Number: 125092-42-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=215edfe243817318d651e48e6eebd5a4f2f0d862&vid=215edfe243817318d651e48e6eebd5a4f2f0d862&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR