Acetamidoxime ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Acetamide oxime

A151385-100G A151385-1G A151385-25G A151385-5G
ALA151385-100GEA 650.66 USD
ALA151385-100G ALA151385-1G ALA151385-25G ALA151385-5G
Acetamidoxime ≥97% (by GC)
Acetamidoxime
Formula: C₂H₆N₂O
MW: 74.08 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00603514
CAS Number: 22059-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d1716cfc6c7411aa005b4ff2d90e290d7bf739f&vid=7d1716cfc6c7411aa005b4ff2d90e290d7bf739f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR