3-Azidopropylamine ≥95% (by GC, titration analysis)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
3-Aminopropyl Azide, 3-Azido-1-propanamine, 3-Azido-1-propylamine, 1-Amino-3-azidopropane
A151380-100MG
A151380-1G
A151380-25MG
A151380-500MG
A151380-5G
ALA151380100MGEA
81.61
USD
ALA151380100MG
ALA151380-1G
ALA151380-25MG
ALA151380500MG
ALA151380-5G
3-Azidopropylamine ≥95% (by GC, titration analysis)
3-Azidopropylamine
3-Azido-1-propanamine can be used to functionalize:
Bismethylolpropionic acid (bis-MPA) monomers with azide functional group to generate high-generation dendrimers.
Clickable zinc tetraphenylporphyrin scaffold with an azido group through click chemistry applicable in photodynamic therapy.
Formula:
C₃H₈N₄ MW: 100.13 g/mol Boiling Pt: 50°C Flash Pt: 60°C Storage Temperature: Freezer |
MDL Number:
MFCD11046568 CAS Number: 88192-19-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...