Azulene ≥97% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Bicyclo[5.3.0]decapentaene
A151375-100MG
A151375-1G
A151375-250MG
A151375-5G
ALA151375100MGEA
66.41
USD
ALA151375100MG
ALA151375-1G
ALA151375250MG
ALA151375-5G
Azulene ≥97% (by GC)
Azulene
Formula:
C₁₀H₈ MW: 128.17 g/mol Boiling Pt: 242 Melting Pt: 98-103°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003810 CAS Number: 275-51-4 Merck Index: 13,00928 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...