1-Adamantanemethylamine ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Adamantan-1-ylmethanamine, 1-Aminomethyladamantane

A151360-100G A151360-1G A151360-250MG A151360-25G A151360-5G
ALA151360-100GEA 5351.26 USD
ALA151360-100G ALA151360-1G ALA151360250MG ALA151360-25G ALA151360-5G
1-Adamantanemethylamine ≥97% (by GC)
1-Adamantanemethylamine

1-Adamantanemethylamine is a large amine present on the wall of 6,7-diaminoquinoxaline. The thermodynamic complex formation constant of 1-adamantanemethylamine was studied.1-Adamantanemethylamine is a large amine present on the wall of 6,7-diaminoquinoxaline. The thermodynamic complex formation constant of 1-adamantanemethylamine was studied.

Formula: C₁₁H₁₉N
MW: 165.28 g/mol
Boiling Pt: 97 °C
Flash Pt: 92°C
Storage Temperature: Ambient
MDL Number: MFCD00074750
CAS Number: 17768-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ef098bfd7fc7d08216c147faef7412095d54598&vid=9ef098bfd7fc7d08216c147faef7412095d54598&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR