4-Acetyldiphenyl sulfide ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151359-1G A151359-5G
ALA151359-1GEA 11.31 USD
ALA151359-1G ALA151359-5G
4-Acetyldiphenyl sulfide ≥98%
4-Acetyldiphenyl sulphide
Formula: C₁₄H₁₂OS
Boiling Pt: 184°C
Melting Pt: 67 °C
MDL Number: MFCD00026227
CAS Number: 10169-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c102e01ac23144f8d35121f3d33a76fe35dbac83&vid=c102e01ac23144f8d35121f3d33a76fe35dbac83&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR