2-Amino-2-ethyl-1,3-propanediol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-2-ethylpropane-1,3-diol

A151350-100G A151350-25G
ALA151350-100GEA 58.81 USD
ALA151350-100G ALA151350-25G
2-Amino-2-ethyl-1,3-propanediol ≥98%
2-Amino-2-ethyl-1,3-propanediol
Formula: C₅H₁₃NO₂
Boiling Pt: 152-153 ℃
Melting Pt: 34°C
Density: 1.099 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00004680
CAS Number: 115-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44cc702aec867b22d902ba80c1e01c6daf17731e&vid=44cc702aec867b22d902ba80c1e01c6daf17731e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR