5-Aminovaleric acid hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 5-Aminovaleric Acid Hydrochloride, 5-Aminopentanoic acid, 5-Aminopentanoic acid Hydrochloride, 5-AVACl, Homopiperidinic acid Hydrochloride, 5-Aminopentanoic acid hydrochloride

A151349-1G A151349-200MG
ALA151349-1GEA 22.71 USD
ALA151349-1G ALA151349200MG
5-Aminovaleric acid hydrochloride ≥98%
5-Aminovaleric acid hydrochloride
Formula: H₂N(CH₂)₄CO₂H·HCl
Melting Pt: 93-97°C
Storage Temperature: Ambient
MDL Number: MFCD00012919
CAS Number: 627-95-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24934e920a03b81c43ceeb89670d77c8dd65e73d&vid=24934e920a03b81c43ceeb89670d77c8dd65e73d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR