4-Aminophenyl 4-Aminobenzoate ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Aminobenzoic acid 4-Aminophenyl ester, 4-Aminophenyl 4-aminobenzoate

A151347-1G A151347-250MG A151347-25G A151347-5G
ALA151347-1GEA 231.71 USD
ALA151347-1G ALA151347250MG ALA151347-25G ALA151347-5G
4-Aminophenyl 4-Aminobenzoate ≥97%
4-Aminophenyl 4-Aminobenzoate
Formula: C₁₃H₁₂N₂O₂
Melting Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD16038468
CAS Number: 20610-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d57ee3977aadc9e0fc56968ba22b931daa0687b&vid=1d57ee3977aadc9e0fc56968ba22b931daa0687b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR