4-Amino-2,3,5,6-tetrafluorobenzamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151346-1G A151346-250MG A151346-25G A151346-5G
ALA151346-1GEA 45.51 USD
ALA151346-1G ALA151346250MG ALA151346-25G ALA151346-5G
4-Amino-2,3,5,6-tetrafluorobenzamide ≥98%
4-Amino-2,3,5,6-tetrafluorobenzamide
Formula: C₇H₄F₄N₂O
Melting Pt: 188 °C
MDL Number: MFCD00008000
CAS Number: 1548-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c44e906acbdfac61e5ece5c1153db73e81e16cb&vid=9c44e906acbdfac61e5ece5c1153db73e81e16cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR