5-Amino-2-methylindole ≥98% (by GC, titration analysis)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Methyl-5-indolamine, 2-Methylindol-5-ylamine, 5-Amino-2-methyl-1H-indole
A151343-1G
A151343-200MG
A151343-25G
A151343-5G
ALA151343-1GEA
30.31
USD
ALA151343-1G
ALA151343200MG
ALA151343-25G
ALA151343-5G
5-Amino-2-methylindole ≥98% (by GC, titration analysis)
5-Amino-2-methylindole
Formula:
C₉H₁₀N₂ MW: 146.19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02093426 CAS Number: 7570-49-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...