4-Allylanisole ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1-Allyl-4-methoxybenzene, Estragole
A151340-100G
A151340-25G
ALA151340-100GEA
72.11
USD
ALA151340-100G
ALA151340-25G
4-Allylanisole ≥98% (by GC)
4-Allylanisole
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 216 Melting Pt: 22.5 Flash Pt: 81 °C |
MDL Number:
MFCD00008653 CAS Number: 140-67-0 Merck Index: 13,03740 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...