Allyl phenyl sulfone ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Phenyl 2-propenyl sulfone, Allyl Phenyl sulfone, Allylsulfonylbenzene, Allyl Phenyl sulphone, (Allylsulphonyl)benzene, (2-Propen-1-ylsulfonyl)benzene, (2-Propenylsulfonyl)benzene, (Allylsulfonyl)benzene

A151331-1G A151331-5G
ALA151331-1GEA 11.31 USD
ALA151331-1G ALA151331-5G
Allyl phenyl sulfone ≥95% (by GC)
Allyl phenyl sulphone
Formula: C₉H₁₀O₂S
Boiling Pt: 128°C
Density: 1.19 g/cm³
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014739
CAS Number: 16212-05-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f27c26ee7c29785f890ce82f4bc086106015431&vid=3f27c26ee7c29785f890ce82f4bc086106015431&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR