2-Amino-5-nitropyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151325-1G A151325-250MG A151325-5G
ALA151325-1GEA 36.01 USD
ALA151325-1G ALA151325250MG ALA151325-5G
2-Amino-5-nitropyrimidine ≥98%
2-Amino-5-nitropyrimidine
Formula: C₄H₄N₄O₂
MW: 140.1
Melting Pt: 235
Density: 1.558
MDL Number: MFCD00006103
CAS Number: 3073-77-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=850e5356ee76abb4a49de4da95ba9991e22dfc23&vid=850e5356ee76abb4a49de4da95ba9991e22dfc23&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR