2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride, 1,2-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)propan-2-yl)diazene dihydrochloride, 2-{2-[(E)-2-[2-(4,5-Dihydro-1H-imidazol-2-yl)proan-2-yl]diazen-1-yl]propan-2-yl}-4,5-dihydro-1H-imidazole dihydrochloride
A151317-100G
A151317-250G
A151317-25G
ALA151317-100GEA
68.31
USD
ALA151317-100G
ALA151317-250G
ALA151317-25G
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥98% (by HPLC)
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride
Formula:
C₁₂H₂₄Cl₂N₆ MW: 323.27 g/mol Melting Pt: 190 °C |
MDL Number:
MFCD00142723 CAS Number: 27776-21-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...