4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4'-[(6-Acryloyloxy)hexyloxy]-4-biphenylcarbonitrile, 6-[(4'-Cyanobiphenyl-4-yl)oxy]hexyl acrylate, Acrylic acid 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl ester, 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl acrylate, 6-((4'-Cyano-[1,1'-biphenyl]-4-yl)oxy)hexyl acrylate

A151308-250MG A151308-25G A151308-5G A151308-1G
ALA151308250MGEA 49.31 USD
ALA151308250MG ALA151308-25G ALA151308-5G ALA151308-1G
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl ≥98%
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl
Formula: C₂₂H₂₃NO₃
MW: 349.43 g/mol
Melting Pt: 71°C
MDL Number: MFCD16293619
CAS Number: 89823-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4659d1c63f37382d80ff51c4dd41bcbd7481837&vid=c4659d1c63f37382d80ff51c4dd41bcbd7481837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR