1,4-Anhydroerythritol ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1,4-Anhydroerythritol, cis-Tetrahydrofuran-3,4-diol, cis-3,4-Tetrahydrofurandiol
A151297-1G
A151297-25G
A151297-5G
ALA151297-1GEA
19.86
USD
ALA151297-1G
ALA151297-25G
ALA151297-5G
1,4-Anhydroerythritol ≥95%
1,4-Anhydroerythritol
Product Description1,4-Anhydroerythritol is a cyclic vicinal diol. 1,4-Anhydroerythritol undergoes hydrodeoxygenation over tungsten oxide-palladium catalysts to yield 3-hydroxytetrahydrofuran.Product Application1,4-Anhydroerythritol was employed in a key step in the synthesis of glucopyranoside-based analogs of adenophostin A lacking the adenine component.
Formula:
C₄H₈O₃ Melting Pt: 19 °C Density: 1.27 g/cm³ Flash Pt: >113 °C |
MDL Number:
MFCD00075210 CAS Number: 4358-64-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...