3a,4,7,7a-Tetrahydroindene ≥98% (by GC) stabilised
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A151294-100ML
A151294-25ML
A151294-50ML
A151294-5ML
ALA151294100MLEA
256.41
USD
ALA151294100ML
ALA151294-25ML
ALA151294-50ML
ALA151294-5ML
3a,4,7,7a-Tetrahydroindene ≥98% (by GC) stabilised
3a,4,7,7a-Tetrahydroindene
Formula:
C₉H₁₂ MW: 120.20 g/mol Boiling Pt: 160°C Flash Pt: 38°C |
MDL Number:
MFCD00021227 CAS Number: 3048-65-5 UN: 3295 ADR: 3 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...