2-[(Acetylthio)methyl]-3-phenylpropionic acid ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-[(Acetylthio)methyl]-3-phenylpropionic Acid

A151283-1G A151283-25G A151283-5G
ALA151283-1GEA 54.06 USD
ALA151283-1G ALA151283-25G ALA151283-5G
2-[(Acetylthio)methyl]-3-phenylpropionic acid ≥98% (by titration)
2-[(Acetylthio)methyl]-3-phenylpropionic acid
Formula: C₁₂H₁₄O₃S
MW: 238.30 g/mol
Melting Pt: 39 °C
MDL Number: MFCD00236751
CAS Number: 91702-98-6
EINECS: 677-137-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2fc02bdef4c07d526334cac0b00b13edc249a9f&vid=d2fc02bdef4c07d526334cac0b00b13edc249a9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR