4-Amino-2,6-dichloropyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2,6-Dichloropyrimidin-4-ylamine

A151268-100G A151268-1G A151268-25G A151268-5G
ALA151268-100GEA 549.01 USD
ALA151268-100G ALA151268-1G ALA151268-25G ALA151268-5G
4-Amino-2,6-dichloropyrimidine ≥98%
4-Amino-2,6-dichloropyrimidine

It is used in the Suzuki coupling of 5-chloro-2-methoxyphenyl boronic acid with 4-amino-2,6-dichloropyrimidine yielded aminochloropyrimidine.

Formula: C₄H₃Cl₂N₃
Melting Pt: 254°C
Storage Temperature: Refrigerator
MDL Number: MFCD00038015
CAS Number: 10132-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7a6f1bcb330d52031ca27a924eca038ccfc634c&vid=b7a6f1bcb330d52031ca27a924eca038ccfc634c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR