2-Amino-m-cresol ≥98% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Hydroxy-6-methylaniline, 2-Amino-3-hydroxytoluene, 2-Amino-3-methylphenol, 1-Amino-2-hydroxy-6-methylbenzene

A151257-100G A151257-1G A151257-25G A151257-5G
ALA151257-100GEA 958.46 USD
ALA151257-100G ALA151257-1G ALA151257-25G ALA151257-5G
2-Amino-m-cresol ≥98% (by GC, titration analysis)
2-Amino-m-cresol

Product Description2-Amino-3-methylphenol is an aminophenol formed by incubating isopropyl-β-D-thiogalactopyranoside-induced E. coli JS996 strain cells with various nitroaromatic compounds.Product Application2-Amino-3-methylphenol was used in the preparation of Schiff base ligands.

Formula: C₇H₉NO
MW: 123.15 g/mol
Melting Pt: 150°C
Storage Temperature: Ambient
MDL Number: MFCD00075082
CAS Number: 2835-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b30329f087c7f037b7af5315f60c5d4136f71678&vid=b30329f087c7f037b7af5315f60c5d4136f71678&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR