5-Amino-8-hydroxyquinoline dihydrochloride ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5-Amino-8-quinolinol Dihydrochloride, 5-Amino-8-hydroxyquinoline Dihydrochloride
A151249-1G
A151249-25G
A151249-5G
ALA151249-1GEA
33.16
USD
ALA151249-1G
ALA151249-25G
ALA151249-5G
5-Amino-8-hydroxyquinoline dihydrochloride ≥98%
5-Amino-8-hydroxyquinoline dihydrochloride
5-Amino-8-hydroxyquinoline dihydrochloride was used in synthesis of: . 5-acetamido-8-hydroxyquinoline hydrochloride . 5-(p-tolylsulfonylimino)quinolin-8-one . 5,8-quinolinesemiquinone via ferric chloride oxidation
Formula:
C₉H₈N₂O Melting Pt: 279°C Storage Temperature: Ambient |
MDL Number:
MFCD00012737 CAS Number: 21302-43-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...