8-Amino-2-methylquinoline ≥97% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
2-Methylquinolin-8-amine, 8-Aminoquinaldine
A151248-100G
A151248-10G
A151248-1G
A151248-250MG
A151248-25G
A151248-5G
ALA151248-100GEA
1987.31
USD
ALA151248-100G
ALA151248-10G
ALA151248-1G
ALA151248250MG
ALA151248-25G
ALA151248-5G
8-Amino-2-methylquinoline ≥97% (by GC)
8-Amino-2-methylquinoline
Formula:
C₁₀H₁₀N₂ Melting Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023998 CAS Number: 18978-78-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...