2,4-Diaminophenol dihydrochloride ≥98% (by total nitrogen and titration analysis)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol
A151245-100G
A151245-1G
A151245-25G
A151245-5G
ALA151245-100GEA
113.91
USD
ALA151245-100G
ALA151245-1G
ALA151245-25G
ALA151245-5G
2,4-Diaminophenol dihydrochloride ≥98% (by total nitrogen and titration analysis)
2,4-Diaminophenol dihydrochloride
Formula:
C₆H₁₀Cl₂N₂O Melting Pt: 205°C |
MDL Number:
MFCD00012979 CAS Number: 137-09-7 UN: 2811 ADR: 6.1,I |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...