6-Amino-5-bromoquinoxaline ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151240-100G A151240-25G A151240-5G
ALA151240-100GEA 1157.01 USD
ALA151240-100G ALA151240-25G ALA151240-5G
6-Amino-5-bromoquinoxaline ≥98%
6-Amino-5-bromoquinoxaline
Formula: C₈H₆BrN₃
MW: 224.0573 g/mol
Melting Pt: 155.0 to 159.0 °C
MDL Number: MFCD00799024
CAS Number: 50358-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e27457672709b3bdca1e0705e95930b01305e16a&vid=e27457672709b3bdca1e0705e95930b01305e16a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR