1-Aminobenzotriazole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1H-Benzo[d][1,2,3]triazol-1-amine

A151238-100MG A151238-10G A151238-1G A151238-250MG A151238-5G
ALA151238100MGEA 55.96 USD
ALA151238100MG ALA151238-10G ALA151238-1G ALA151238250MG ALA151238-5G
1-Aminobenzotriazole ≥98%
1-Aminobenzotriazole

1-Aminobenzotriazole (ABT) is a non-specific cytochrome P450 (CYP) inhibitor.

Formula: C₆H₆N₄
MW: 134.14 g/mol
Melting Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00132902
CAS Number: 1614-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c76c31bb1c8daf23bb20c8c854bf9a40ecbff34&vid=9c76c31bb1c8daf23bb20c8c854bf9a40ecbff34&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR