Acetaldehyde semicarbazone ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Acetaldehyde Semicarbazone

A151236-1G A151236-25G A151236-5G
ALA151236-1GEA 15.11 USD
ALA151236-1G ALA151236-25G ALA151236-5G
Acetaldehyde semicarbazone ≥98%
Acetaldehyde semicarbazone
Formula: C₃H₇N₃O
Melting Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00059191
CAS Number: 591-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e952d520ddbb4a16cb7fab6bcc747c7283f7f110&vid=e952d520ddbb4a16cb7fab6bcc747c7283f7f110&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR