3-Acetyl-2,4-dimethylpyrrole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151235-1G A151235-5G
ALA151235-1GEA 40.76 USD
ALA151235-1G ALA151235-5G
3-Acetyl-2,4-dimethylpyrrole ≥98%
3-Acetyl-2,4-dimethylpyrrole
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 173°C
Melting Pt: 136-140°C
MDL Number: MFCD00005221
CAS Number: 2386-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab95a4b82bea41bfa56364ff70a4ccdd254c78a7&vid=ab95a4b82bea41bfa56364ff70a4ccdd254c78a7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR