3'-Aminoacetanilide ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: N-(3-Aminophenyl)acetamide, 3-Acetamidoaniline, N-Acetyl-1,3-phenylenediamine, m-Aminoacetanilide

A151227-100G A151227-250G
ALA151227-100GEA 96.81 USD
ALA151227-100G ALA151227-250G
3'-Aminoacetanilide ≥98% (by HPLC)
3'-Aminoacetanilide
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Boiling Pt: 388
Melting Pt: 85-89°C
MDL Number: MFCD00025229
CAS Number: 102-28-3
EINECS: 203-021-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=677b4946ed759683356e8366c6f33ee5415b5ca7&vid=677b4946ed759683356e8366c6f33ee5415b5ca7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR