ɑ,ɑ-Dimethyl-γ-butyrolactone ≥97% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Dihydro-3,3-dimethyl-2(3H)-furanone, α,α-Dimethyl-γ-butyrolactone
A151224-1G
A151224-5G
ALA151224-1GEA
97.76
USD
ALA151224-1G
ALA151224-5G
ɑ,ɑ-Dimethyl-γ-butyrolactone ≥97% (by GC)
alpha,alpha-Dimethyl-gamma-butyrolactone
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 194°C Melting Pt: 6°C Flash Pt: 67 °C |
MDL Number:
MFCD03844788 CAS Number: 3709-08-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...