3-Acetylthiazolidine-2-thione ≥98%

Supplier: ALADDIN SCIENTIFIC

A151215-1G A151215-25G A151215-5G
ALA151215-1GEA 35.06 USD
ALA151215-1G ALA151215-25G ALA151215-5G
3-Acetylthiazolidine-2-thione ≥98%
3-Acetylthiazolidine-2-thione
Formula: C₅H₇NOS₂
Flash Pt: 104 °C
Storage Temperature: Ambient
CAS Number: 76397-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d8fa06f9a163467a00ef6db7ee108e55ff8068a&vid=4d8fa06f9a163467a00ef6db7ee108e55ff8068a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR