2-Acetamidothiazole ≥98%

Supplier: ALADDIN SCIENTIFIC

A151213-100G A151213-25G A151213-5G
ALA151213-100GEA 306.76 USD
ALA151213-100G ALA151213-25G ALA151213-5G
2-Acetamidothiazole ≥98%
2-Acetamidothiazole
Formula: C₅H₆N₂OS
Melting Pt: 202-209°C
MDL Number: MFCD00016892
CAS Number: 2719-23-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65cd53b33871a05d7c5c3f186c80951cf68455c1&vid=65cd53b33871a05d7c5c3f186c80951cf68455c1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR