2-Aminobenzothiazole hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Aminobenzothiazole Hydrochloride

A151210-25G A151210-5G
ALA151210-25GEA 76.86 USD
ALA151210-25G ALA151210-5G
2-Aminobenzothiazole hydrochloride ≥97%
2-Aminobenzothiazole hydrochloride
Formula: C₇H₆N₂S
Melting Pt: 240 °C
MDL Number: MFCD00142859
CAS Number: 94787-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf676f0f9f1e76bee31624d8e6e9abab79f1bcb4&vid=bf676f0f9f1e76bee31624d8e6e9abab79f1bcb4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR