2-Amino-5-nitrothiazole ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine

A151201-100G A151201-25G
ALA151201-100GEA 219.36 USD
ALA151201-100G ALA151201-25G
2-Amino-5-nitrothiazole ≥98% (by HPLC)
2-Amino-5-nitrothiazole

2-Amino-5-nitrothiazole(ANT) is a hypoxic radiosensitizing drug. ANT forms square-planar complex, trans-[PdCl2(ANT)2] with palladium in methanol2-Amino-5-nitrothiazole was used as diazo component in the synthesis of monoazo disperse dyes. It was used as matrix during matrix-assisted laser desorption/ionization time-of-flight mass spectrometric study of oligonucleotide and protein.

Formula: C₃H₃N₃O₂S
MW: 145.14 g/mol
Melting Pt: 203
MDL Number: MFCD00005326
CAS Number: 121-66-4
EINECS: 204-490-9
UN: 2811
ADR: 6.1 I
Merck Index: 13,00456
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1739f17363abbba40e13e634f0c350be0175c9a0&vid=1739f17363abbba40e13e634f0c350be0175c9a0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR