N-Allylrhodanine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151200-25G A151200-5G
ALA151200-25GEA 227.91 USD
ALA151200-25G ALA151200-5G
N-Allylrhodanine ≥98% (by GC)
N-Allylrhodanine
Formula: C₆H₇NOS₂
Boiling Pt: 139 °C
Melting Pt: 46.0to49.0°C
Density: 1.135 g/cm³
MDL Number: MFCD00044700
CAS Number: 1457-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d70491a4bb4cb998b8a742808ee877c0bbbde53&vid=6d70491a4bb4cb998b8a742808ee877c0bbbde53&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR