2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151199-250MG A151199-1G
ALA151199250MGEA 11.31 USD
ALA151199250MG ALA151199-1G
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98%
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Formula: C₁₁H₁₀ClN₃OS
MW: 267.73 g/mol
Melting Pt: 210 °C
MDL Number: MFCD10000630
CAS Number: 302964-24-5
EINECS: 460-280-9, 608-455-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d579e69aa4b5c7967b72f12f5359e0fcfed0b321&vid=d579e69aa4b5c7967b72f12f5359e0fcfed0b321&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR