2-Amino-4-chlorobenzothiazole ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151196-100G A151196-1G A151196-25G A151196-5G
ALA151196-100GEA 468.26 USD
ALA151196-100G ALA151196-1G ALA151196-25G ALA151196-5G
2-Amino-4-chlorobenzothiazole ≥98% (by HPLC)
2-Amino-4-chlorobenzothiazole
Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00005791
CAS Number: 19952-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=831060c794a94215463535b4b9d8fcb70c3a71fb&vid=831060c794a94215463535b4b9d8fcb70c3a71fb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR