ɑ-Cyano-4-hydroxycinnamic acid ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Cyano-3-(4-hydroxyphenyl)acrylic Acid, α-Cyano-4-hydroxycinnamic Acid, α-CHCA, CHCA

A151195-100G A151195-1G A151195-25G A151195-5G
ALA151195-100GEA 5987.76 USD
ALA151195-100G ALA151195-1G ALA151195-25G ALA151195-5G
ɑ-Cyano-4-hydroxycinnamic acid ≥98%, Moligand™
alpha-Cyano-4-hydroxycinnamic acid

Product Introduction:
α-Cyano-4-hydroxycinnamic acid is the commonly used matrix for analyzing peptides by matrix-assisted laser desorption/ionization mass spectrometry. It inhibits the monophenolase activity and diphenolase activity of mushroom tyrosinase.Applicationα-Cyano-4-hydroxycinnamic acid (CHC) was used in encapsulation of CHC into NaY zeolite. It was used as matrix to investigate the activity of the paclitaxel derivatives using several well-established in vitro angiogenesis assays.

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 245-250°C
Storage Temperature: Refrigerator
MDL Number: MFCD00004204
CAS Number: 28166-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75610f07e3c30baead63b8f6950ab688534fbd33&vid=75610f07e3c30baead63b8f6950ab688534fbd33&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR