5-Azidovaleric acid ≥95%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 5-Azidopentanoic acid, 5-Azidovaleric Acid, 5-Azidopentanoic Acid

A151193-250MG A151193-50MG
ALA151193250MGEA 848.26 USD
ALA151193250MG ALA151193-50MG
5-Azidovaleric acid ≥95%
5-Azidovaleric acid
Formula: C₅H₉N₃O₂
Boiling Pt: 85°C
Density: 1.14 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD11850108
CAS Number: 79583-98-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82b8f842d5d0ce8b932dde82b62e6c4e52be43f&vid=f82b8f842d5d0ce8b932dde82b62e6c4e52be43f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR