Amezinium methyl sulfate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-6-methoxy-1-phenylpyridazinium Methyl sulfate, 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate, Amezinium Methyl sulphate, 4-Amino-6-methoxy-1-phenylpyridazin-1-ium methyl sulfate, Lu-1631, 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulphate, Amezinium methylsulphate, Amezinium methylsulfate, Amezinium metilsulfate, 4-Amino-6-methoxy-1-phenylpyridazin-1-ium methyl sulphate, Amezinium Methyl sulfate

A151192-1G A151192-250MG
ALA151192-1GEA 36.96 USD
ALA151192-1G ALA151192250MG
Amezinium methyl sulfate ≥98%
Amezinium methyl sulfate

Product Introduction:Amezinium metalsulfonate stimulates alpha and beta-1 receptors, inhibiting the absorption of norepinephrine and tyramine.

Formula: C₁₂H₁₅N₃O₅S
Melting Pt: 176°C
Storage Temperature: Freezer
MDL Number: MFCD00865972
CAS Number: 30578-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead4450b19a44397fda5993f827213d33eb950bc&vid=ead4450b19a44397fda5993f827213d33eb950bc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR