Aberchrome 670 ≥95%

Supplier: ALADDIN SCIENTIFIC

Synonyms: (E)-3-(Adamantan-2-ylidene)-4-[1-(2,5-dimethyl-3-furyl)ethylidene]dihydro-2,5-furandione

A151189-1G A151189-200MG A151189-50MG
ALA151189-1GEA 527.16 USD
ALA151189-1G ALA151189200MG ALA151189-50MG
Aberchrome 670 ≥95%
Aberchrome 670

Aberchrom 670 is a surface active agent that can be used in the production of coatings and other devices. It has a high solubility parameter, which means it has a low surface tension and can form films on water surfaces. Aberchrom 670 is an amphiphilic molecule, which means it has both hydrophobic and hydrophilic parts. This property allows for the formation of films on water surfaces, making it useful in applications such as coating and device manufacturing. The phase transition temperature of Aberchrom 670 is higher than room temperature, so it will not melt when exposed to UV radiation. Aberchrom 670 also fluoresces under UV irradiation, allowing for easy detection during manufacturing processes.

Formula: C₂₂H₂₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD07368579
CAS Number: 94856-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92e6ccbf3f060b73d80e5d3afa5144a76d72c617&vid=92e6ccbf3f060b73d80e5d3afa5144a76d72c617&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR