1-Adamantaneethanol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-(2-Hydroxyethyl)adamantane, 1-(2-Hydroxyethyl)tricyclo[3.3.1.1(3,7)]decane, 2-(1-Adamantyl)ethanol

A151184-1G A151184-5G
ALA151184-1GEA 25.56 USD
ALA151184-1G ALA151184-5G
1-Adamantaneethanol ≥98% (by GC)
1-Adamantaneethanol

1-Adamantaneethanol was used in the synthesis of:4-(adamant-1-ylethylenoxycarbonyl)phthalanhydridepoly(2-methoxy-5-cyclohexylmethyloxy-p-phenylene vinylene)adamantine-containing phthalocyanines

Formula: C₁₂H₂₀O
Melting Pt: 66-78°C
MDL Number: MFCD00074756
CAS Number: 6240-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR