1-Acetyladamantane ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151183-100G A151183-1G A151183-25G A151183-5G
ALA151183-100GEA 656.36 USD
ALA151183-100G ALA151183-1G ALA151183-25G ALA151183-5G
1-Acetyladamantane ≥98% (by GC)
1-Acetyladamantane

Product Description1-Adamantyl methyl ketone, a sterically bulky ketone substrate, was reduced to the corresponding alcohol with excellent optical purity.Product Application1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Formula: C₁₂H₁₈O
Melting Pt: 54 °C
Flash Pt: 108 °C
MDL Number: MFCD00074739
CAS Number: 1660-04-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6df39481c63b137b581f3efc9d999111c912583&vid=d6df39481c63b137b581f3efc9d999111c912583&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR