4-(1-Adamantyl)phenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151181-100G A151181-1G A151181-250MG A151181-25G A151181-5G
ALA151181-100GEA 2749.21 USD
ALA151181-100G ALA151181-1G ALA151181250MG ALA151181-25G ALA151181-5G
4-(1-Adamantyl)phenol ≥98%
4-(1-Adamantyl)phenol
Formula: C₁₆H₂₀O
Melting Pt: 179 °C
Storage Temperature: Freezer
MDL Number: MFCD00168143
CAS Number: 29799-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2097b2bd566cee798b529ad1968d58287c5b76ac&vid=2097b2bd566cee798b529ad1968d58287c5b76ac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR